Please login to your online account to display your discounted pricing

Rat anti-TER-119, Clone: TER-119, PE-Cy5, Novus Biologicals™

$321.00 - $418.00

Specifications

Antigen TER-119
Clone TER-119
Concentration 0.2mg/mL
Immunogen Day-14 fetal liver cells from a C57BL/6 mouse
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20036610

 
novus biologicals canada
NBP200366100UG
100μg Each for $321.00

NBP20036626

 
novus biologicals canada
NBP200366200UG
200μg Each for $418.00
Description & Specifications

Specifications

Antigen TER-119
Clone TER-119
Concentration 0.2mg/mL
Immunogen Day-14 fetal liver cells from a C57BL/6 mouse
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate PE-Cy5
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry