Please login to your online account to display your discounted pricing

Rat anti-TER-119, Clone: TER-119, PE-Cy5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200366200UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20036626

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Antigen TER-119
Format Affinity Purified
Immunogen Day-14 fetal liver cells from a C57BL/6 mouse
Isotype IgG2b, κ
Purification Method Affinity Purified
Quantity 200μg
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone TER-119
Conjugate PE-Cy5
Host Species Rat
Storage Requirements Store at 4°C in the dark.
Test Specificity TER-119 (TER-119)
Regulatory Status RUO
Concentration 0.2mg/mL
Cross Reactivity Mouse