Please login to your online account to display your discounted pricing

Rat anti-TER-119, Clone: TER-119, PE-Cy5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200366100UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20036610

  • / Each
 

Description & Specifications

Specifications

Immunogen Day-14 fetal liver cells from a C57BL/6 mouse
Primary or Secondary Primary
Conjugate PE-Cy5
Isotype IgG2b, κ
Applications Flow Cytometry
Format Affinity Purified
Clone TER-119
Cross Reactivity Mouse
Host Species Rat
Quantity 100μg
Regulatory Status RUO
Antigen TER-119
Monoclonal or Polyclonal Monoclonal
Test Specificity TER-119 (TER-119)
Concentration 0.2mg/mL
Purification Method Affinity Purified
Storage Requirements Store at 4°C in the dark.