Please login to your online account to display your discounted pricing

Rat anti-TCR V beta 8.3, Clone: B21.14, PE, Novus Biologicals™

$362.00 - $505.00

Specifications

Antigen TCR V beta 8.3
Clone B21.14
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody is TCR V beta 8.3.
Gene ID (Entrez) 6957
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20013410

 
novus biologicals canada
NBP200134100UG
100μg Each for $362.00

NBP20013426

 
novus biologicals canada
NBP200134200UG
200μg Each for $505.00
Description & Specifications

Specifications

Antigen TCR V beta 8.3
Clone B21.14
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody is TCR V beta 8.3.
Gene ID (Entrez) 6957
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry