Please login to your online account to display your discounted pricing

Rat anti-RANK/TNFRSF11A, Clone: R12-31, Biotin, Novus Biologicals™

$192.00 - $493.00

Specifications

Antigen RANK/TNFRSF11A
Clone R12-31
Concentration 0.5mg/mL
Immunogen The immunogen was the full length version of the protein: RANK (R12-31)
Gene ID (Entrez) 8792
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14349501

 
novus biologicals canada
NBP14349501MG
0.1mg Each for $246.00

NBP1434955

 
novus biologicals canada
NBP14349505MG
0.5mg Each for $493.00

NBP14349505

 
novus biologicals canada
NBP143495005MG
0.05mg Each for $192.00
Description & Specifications

Specifications

Antigen RANK/TNFRSF11A
Clone R12-31
Concentration 0.5mg/mL
Immunogen The immunogen was the full length version of the protein: RANK (R12-31)
Gene ID (Entrez) 8792
Formulation PBS [pH 7.2]. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry