Please login to your online account to display your discounted pricing

Rat anti-RANK/TNFRSF11A, Clone: R12-31, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14349505MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1434955

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Formulation PBS [pH 7.2]. Immunogen affinity purified.
Immunogen The immunogen was the full length version of the protein: RANK (R12-31)
Format Affinity Purified
Gene ID (Entrez) 8792
Primary or Secondary Primary
Conjugate Biotin
Cross Reactivity Mouse
Storage Requirements Store at 4°C. Do not freeze.
Regulatory Status RUO
Concentration 0.5mg/mL
Antigen RANK/TNFRSF11A
Clone R12-31
Monoclonal or Polyclonal Monoclonal
Quantity 0.5mg
Isotype IgG2a, κ
Test Specificity RANK (R12-31)
Purification Method Affinity Purified
Host Species Rat