Please login to your online account to display your discounted pricing

Rat anti-RANK/TNFRSF11A, Clone: R12-31, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14349501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14349501

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Antigen RANK/TNFRSF11A
Host Species Rat
Applications Flow Cytometry
Isotype IgG2a, κ
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 8792
Test Specificity RANK (R12-31)
Clone R12-31
Concentration 0.5mg/mL
Format Affinity Purified
Formulation PBS [pH 7.2]. Immunogen affinity purified.
Immunogen The immunogen was the full length version of the protein: RANK (R12-31)
Purification Method Affinity Purified
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Conjugate Biotin