Please login to your online account to display your discounted pricing

Rat anti-RAE1, Clone: CX1, Biotin, Novus Biologicals™

$246.00 - $708.00

Specifications

Antigen RAE1
Clone CX1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was RAE1g.
Gene ID (Entrez) 8480
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14353201

 
novus biologicals canada
NBP14353201MG
0.1mg Each for $321.00

NBP1435325

 
novus biologicals canada
NBP14353205MG
0.5mg Each for $708.00

NBP14353205

 
novus biologicals canada
NBP143532005MG
0.05mg Each for $246.00
Description & Specifications

Specifications

Antigen RAE1
Clone CX1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was RAE1g.
Gene ID (Entrez) 8480
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry