Please login to your online account to display your discounted pricing

Rat anti-PDGF R beta, Clone: APB5, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen PDGF R beta
Clone APB5
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was PDGF receptor beta.
Gene ID (Entrez) 5159
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14334905

 
novus biologicals canada
NBP143349005MG
0.05mg Each for $219.00

NBP14334901

 
novus biologicals canada
NBP14334901MG
0.1mg Each for $285.00
Description & Specifications

Specifications

Antigen PDGF R beta
Clone APB5
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was PDGF receptor beta.
Gene ID (Entrez) 5159
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)