missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-PDGF R alpha, Clone: APA5, Biotin, Novus Biologicals™

$175.00 - $329.00

Specifications

Antigen PDGF R alpha
Clone APA5
Immunogen The immunogen for this antibody was CD140a.
Gene ID (Entrez) 5156
Formulation Phosphate buffer pH 7.2
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14358101 Novus Biologicals Canada
NBP14358101MG
0.1mg Each for $329.00
NBP14358103 Novus Biologicals Canada
NBP1435810025MG
0.025mg Each for $175.00
Specifications Description & Specifications

Specifications

PDGF R alpha
The immunogen for this antibody was CD140a.
Phosphate buffer pH 7.2
Primary
Rat
Biotin
Affinity Purified
APA5
5156
Monoclonal
Store at 4°C. Do not freeze.
IgG2a, κ
Affinity Purified
Flow Cytometry