missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-PDE3B, Clone: SMCP3B, Novus Biologicals™

$307.00 - $428.00

Specifications

Antigen PDE3B
Clone SMCP3B
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was PDE3B.
Gene ID (Entrez) 5140
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14333301 Novus Biologicals Canada
NBP14333301MG
0.1mg Each for $428.00
NBP14333303 Novus Biologicals Canada
NBP1433330025MG
0.025mg Each for $307.00
Specifications Description & Specifications

Specifications

PDE3B
0.5mg/mL
5140
Monoclonal
Store at 4°C. Do not freeze.
IgG2a, κ
Affinity Purified
Western Blot
SMCP3B
The immunogen for this antibody was PDE3B.
Aqueous buffer (contains stabilizer if necessary)
Primary
Rat
Unlabeled
Affinity Purified