Please login to your online account to display your discounted pricing

Rat anti-PDE3B, Clone: SMCP3B, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1433330025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14333303

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Host Species Rat
Immunogen The immunogen for this antibody was PDE3B.
Conjugate Unlabeled
Regulatory Status RUO
Gene ID (Entrez) 5140
Concentration 0.5mg/mL
Purification Method Affinity Purified
Monoclonal or Polyclonal Monoclonal
Applications Western Blotting
Quantity 0.025mg
Formulation Aqueous buffer (contains stabilizer if necessary)
Clone SMCP3B
Storage Requirements Store at 4°C. Do not freeze.
Format Affinity Purified
Primary or Secondary Primary
Isotype IgG2a, κ
Antigen PDE3B