Please login to your online account to display your discounted pricing

Rat anti-PD-L2/B7-DC/PDCD1LG2, Clone: TY25, PE, Novus Biologicals™

$354.00 - $493.00

Specifications

Antigen PD-L2/B7-DC/PDCD1LG2
Clone TY25
Concentration 0.2mg/mL
Immunogen B7-DC transfected RAW264.7 cells.
Gene ID (Entrez) 80380
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20045010

 
novus biologicals canada
NBP200450100UG
100μg Each for $354.00

NBP20045026

 
novus biologicals canada
NBP200450200UG
200μg Each for $493.00
Description & Specifications

Specifications

Antigen PD-L2/B7-DC/PDCD1LG2
Clone TY25
Concentration 0.2mg/mL
Immunogen B7-DC transfected RAW264.7 cells.
Gene ID (Entrez) 80380
Formulation Protein G purified
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a
Conjugate PE
Format Purified
Purification Method Purified
Applications Flow Cytometry