Please login to your online account to display your discounted pricing

Rat anti-PD-L2/B7-DC/PDCD1LG2, Clone: TY25, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200450100UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20045010

  • / Each
 

Description & Specifications

Specifications

Antigen PD-L2/B7-DC/PDCD1LG2
Applications Flow Cytometry
Clone TY25
Concentration 0.2mg/mL
Conjugate PE
Cross Reactivity Mouse
Format Purified
Formulation Protein G purified
Host Species Rat
Immunogen B7-DC transfected RAW264.7 cells.
Isotype IgG2a
Purification Method Purified
Quantity 100μg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 80380
Test Specificity NB100-65332 recognizes mouse CD273, a 42kD member of the B7 family, otherwise known as B7-dendritic cell (B7-DC), or programmed death ligand-2 (PD-L2). The expression of CD273 is restricted to a subpopulation of dendritic cells (DCs), and DCs and macrophages that have been stimulated with interferon gamma, GM-CSF or IL-4. CD273 has been identified as a ligand for PD-1(CD279), along with another B7 family member B7-H1 (CD274). Studies suggest that both CD273 and B7-H1 co-stimulate or inhibit T cell proliferation and cytokine production. Antibodies produced by clone TY25 are reported to block the binding of CD273 (B7-DC) to it's receptor.