Please login to your online account to display your discounted pricing

Rat anti-PD-L2/B7-DC/PDCD1LG2, Clone: TY25, Biotin, Novus Biologicals™

$329.00 - $329.00

Specifications

Antigen PD-L2/B7-DC/PDCD1LG2
Clone TY25
Concentration 0.5mg/mL
Immunogen B7-DC transfected RAW264.7 cells.
Gene ID (Entrez) 80380
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14350401

 
novus biologicals canada
NBP14350401MG
0.1mg Each for $329.00
Description & Specifications

Specifications

Antigen PD-L2/B7-DC/PDCD1LG2
Clone TY25
Concentration 0.5mg/mL
Immunogen B7-DC transfected RAW264.7 cells.
Gene ID (Entrez) 80380
Formulation Phosphate buffer pH 7.2. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2a
Conjugate Biotin
Format Purified
Purification Method Purified
Applications Flow Cytometry