Please login to your online account to display your discounted pricing

Rat anti-PD-L2/B7-DC/PDCD1LG2, Clone: 122, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14311605MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1431165

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Quantity 0.5mg
Immunogen The immunogen for this antibody was B7DC.
Isotype IgG2a, κ
Monoclonal or Polyclonal Monoclonal
Applications Flow Cytometry
Host Species Rat
Cross Reactivity Mouse
Formulation Phosphate buffer pH 7.2
Antigen PD-L2/B7-DC/PDCD1LG2
Storage Requirements Store at 4°C. Do not freeze.
Clone 122
Conjugate Unlabeled
Primary or Secondary Primary
Regulatory Status RUO
Purification Method Affinity Purified
Concentration 0.5mg/mL
Gene ID (Entrez) 80380