Please login to your online account to display your discounted pricing

Rat anti-PD-L2/B7-DC/PDCD1LG2, Clone: 122, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14311601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14311601

  • / Each
 

Description & Specifications

Specifications

Antigen PD-L2/B7-DC/PDCD1LG2
Applications Flow Cytometry
Clone 122
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Rat
Immunogen The immunogen for this antibody was B7DC.
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 80380
Target Species Murine