Please login to your online account to display your discounted pricing

Rat anti-PD-L2/B7-DC/PDCD1LG2, Clone: 122, FITC, Novus Biologicals™

$252.00 - $725.00

Specifications

Antigen PD-L2/B7-DC/PDCD1LG2
Clone 122
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was B7DC.
Gene ID (Entrez) 80380
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14391401

 
novus biologicals canada
NBP14391401MG
0.1mg Each for $329.00

NBP14391405

 
novus biologicals canada
NBP143914005MG
0.05mg Each for $252.00

NBP1439145

 
novus biologicals canada
NBP14391405MG
0.5mg Each for $725.00
Description & Specifications

Specifications

Antigen PD-L2/B7-DC/PDCD1LG2
Clone 122
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was B7DC.
Gene ID (Entrez) 80380
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry