Please login to your online account to display your discounted pricing

Rat anti-PD-1, Clone: RMP1-30, PE, Novus Biologicals™

$362.00 - $505.00

Specifications

Antigen PD-1
Clone RMP1-30
Concentration 0.2mg/mL
Immunogen Mouse PD-1 transfected BHK cells
Gene ID (Entrez) 5133
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20043710

 
novus biologicals canada
NBP200437100UG
100μg Each for $362.00

NBP20043726

 
novus biologicals canada
NBP200437200UG
200μg Each for $505.00
Description & Specifications

Specifications

Antigen PD-1
Clone RMP1-30
Concentration 0.2mg/mL
Immunogen Mouse PD-1 transfected BHK cells
Gene ID (Entrez) 5133
Formulation Immunogen affinity purified
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry