Please login to your online account to display your discounted pricing

Rat anti-PD-1, Clone: RMP1-30, Biotin, Novus Biologicals™

$252.00 - $725.00

Specifications

Antigen PD-1
Clone RMP1-30
Immunogen Mouse PD-1 transfected BHK cells
Gene ID (Entrez) 5133
Formulation Phosphate buffer pH 7.2. Immunogen affinity purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14346501

 
novus biologicals canada
NBP14346501MG
0.1mg Each for $329.00

NBP14346505

 
novus biologicals canada
NBP143465005MG
0.05mg Each for $252.00

NBP1434655

 
novus biologicals canada
NBP14346505MG
0.5mg Each for $725.00
Description & Specifications

Specifications

Antigen PD-1
Clone RMP1-30
Immunogen Mouse PD-1 transfected BHK cells
Gene ID (Entrez) 5133
Formulation Phosphate buffer pH 7.2. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry