Please login to your online account to display your discounted pricing

Rat anti-OX40 Ligand/TNFSF4, Clone: RM134L, PE, Novus Biologicals™

$296.00 - $428.00

Specifications

Antigen OX40 Ligand/TNFSF4
Clone RM134L
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody is TNFSF4.
Gene ID (Entrez) 7292
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20036310

 
novus biologicals canada
NBP200363100UG
100μg Each for $296.00

NBP20036326

 
novus biologicals canada
NBP200363200UG
200μg Each for $428.00
Description & Specifications

Specifications

Antigen OX40 Ligand/TNFSF4
Clone RM134L
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody is TNFSF4.
Gene ID (Entrez) 7292
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay