Please login to your online account to display your discounted pricing

Rat anti-NKG2D/CD314, Clone: MI-6, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP14328805MG

 View all options for this product


For Research Use Only

Catalog No. NBP1432885


Description & Specifications


Antigen NKG2D/CD314
Applications Flow Cytometry
Applications Functional Assay
Clone MI-6
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Rat
Immunogen The immunogen for this antibody was NKG2D CD314.
Isotype IgG2a, λ
Purification Method Affinity Purified
Quantity 0.5mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 22914