Please login to your online account to display your discounted pricing

Rat anti-NKG2D/CD314, Clone: MI-6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143288005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14328805

  • / Each
 

Description & Specifications

Specifications

Host Species Rat
Regulatory Status RUO
Immunogen The immunogen for this antibody was NKG2D CD314.
Cross Reactivity Mouse
Quantity 0.05mg
Antigen NKG2D/CD314
Applications Flow Cytometry
Applications Functional Assay
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Gene ID (Entrez) 22914
Format Affinity Purified
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Clone MI-6
Isotype IgG2a, λ
Formulation Phosphate buffer pH 7.2
Concentration 0.5mg/mL