Please login to your online account to display your discounted pricing

Rat anti-NKG2D/CD314, Clone: MI-6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14328801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14328801

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Host Species Rat
Applications Flow Cytometry
Applications Functional Assay
Immunogen The immunogen for this antibody was NKG2D CD314.
Isotype IgG2a, λ
Quantity 0.1mg
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Antigen NKG2D/CD314
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 22914
Clone MI-6
Concentration 0.5mg/mL
Conjugate Unlabeled
Formulation Phosphate buffer pH 7.2
Cross Reactivity Mouse