Please login to your online account to display your discounted pricing

Rat anti-NKG2D/CD314, Clone: MI-6, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14353301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14353301

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Applications Western Blotting
Antigen NKG2D/CD314
Host Species Rat
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Immunogen The immunogen for this antibody was NKG2D CD314.
Isotype IgG2a, λ
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 22914
Clone MI-6
Concentration 0.5mg/mL
Conjugate Biotin
Cross Reactivity Mouse