Please login to your online account to display your discounted pricing

Rat anti-NKG2D/CD314, Clone: MI-6, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143533005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14353305

  • / Each
 

Description & Specifications

Specifications

Host Species Rat
Applications Flow Cytometry
Applications Functional Assay
Applications Western Blotting
Gene ID (Entrez) 22914
Antigen NKG2D/CD314
Formulation Phosphate buffer pH 7.2
Immunogen The immunogen for this antibody was NKG2D CD314.
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone MI-6
Format Affinity Purified
Isotype IgG2a, λ
Concentration 0.5mg/mL
Conjugate Biotin
Cross Reactivity Mouse