Please login to your online account to display your discounted pricing

Rat anti-NKG2D/CD314, Clone: CX5, PE, Novus Biologicals™

$128.00 - $418.00

Specifications

Antigen NKG2D/CD314
Clone CX5
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was NKG2D CD314.
Gene ID (Entrez) 22914
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14375201

 
novus biologicals canada
NBP143752001MG
0.01mg Each for $128.00

NBP14375202

 
novus biologicals canada
NBP14375202MG
0.2mg Each for $418.00

NBP14375205

 
novus biologicals canada
NBP143752005MG
0.05mg Each for $192.00

NBP1437521

 
novus biologicals canada
NBP14375201MG
0.1mg Each for $289.00
Description & Specifications

Specifications

Antigen NKG2D/CD314
Clone CX5
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was NKG2D CD314.
Gene ID (Entrez) 22914
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry