Please login to your online account to display your discounted pricing

Rat anti-NKG2D/CD314, Clone: CX5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143286005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14328605

  • / Each
 

Description & Specifications

Specifications

Host Species Rat
Immunogen The immunogen for this antibody was NKG2D CD314.
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Formulation Phosphate buffer pH 7.2
Regulatory Status RUO
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay
Clone CX5
Cross Reactivity Mouse
Antigen NKG2D/CD314
Gene ID (Entrez) 22914
Format Affinity Purified
Primary or Secondary Primary
Isotype IgG1, κ
Concentration 0.5mg/mL
Storage Requirements Store at 4°C. Do not freeze.
Quantity 0.05mg