Please login to your online account to display your discounted pricing

Rat anti-NKG2D/CD314, Clone: CX5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14328601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14328601

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Conjugate Unlabeled
Primary or Secondary Primary
Gene ID (Entrez) 22914
Format Affinity Purified
Cross Reactivity Mouse
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Isotype IgG1, κ
Regulatory Status RUO
Formulation Phosphate buffer pH 7.2
Immunogen The immunogen for this antibody was NKG2D CD314.
Clone CX5
Host Species Rat
Storage Requirements Store at 4°C. Do not freeze.
Quantity 0.1mg
Purification Method Affinity Purified
Antigen NKG2D/CD314