Please login to your online account to display your discounted pricing

Rat anti-NKG2D/CD314, Clone: CX5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14328605MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1432865

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Cross Reactivity Mouse
Conjugate Unlabeled
Formulation Phosphate buffer pH 7.2
Format Affinity Purified
Immunogen The immunogen for this antibody was NKG2D CD314.
Gene ID (Entrez) 22914
Antigen NKG2D/CD314
Regulatory Status RUO
Clone CX5
Concentration 0.5mg/mL
Purification Method Affinity Purified
Isotype IgG1, κ
Host Species Rat
Quantity 0.5mg
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal