Please login to your online account to display your discounted pricing

Rat anti-NKG2D/CD314, Clone: CX5, Biotin, Novus Biologicals™

$171.00 - $493.00

Specifications

Antigen NKG2D/CD314
Clone CX5
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was NKG2D CD314.
Gene ID (Entrez) 22914
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14353101

 
novus biologicals canada
NBP14353101MG
0.1mg Each for $246.00

NBP14353105

 
novus biologicals canada
NBP143531005MG
0.05mg Each for $171.00

NBP1435315

 
novus biologicals canada
NBP14353105MG
0.5mg Each for $493.00
Description & Specifications

Specifications

Antigen NKG2D/CD314
Clone CX5
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was NKG2D CD314.
Gene ID (Entrez) 22914
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG1, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry