Please login to your online account to display your discounted pricing

Rat anti-NKG2D/CD314, Clone: CX5, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14353105MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1435315

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Format Affinity Purified
Immunogen The immunogen for this antibody was NKG2D CD314.
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 22914
Antigen NKG2D/CD314
Formulation Phosphate buffer pH 7.2
Host Species Rat
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 0.5mg
Storage Requirements Store at 4°C. Do not freeze.
Clone CX5
Cross Reactivity Mouse
Concentration 0.5mg/mL
Conjugate Biotin