Please login to your online account to display your discounted pricing

Rat anti-NKG2D/CD314, Clone: CX5, Biotin, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP143531005MG

 View all options for this product


For Research Use Only

Catalog No. NBP14353105


Description & Specifications


Antigen NKG2D/CD314
Applications Flow Cytometry
Clone CX5
Concentration 0.5mg/mL
Conjugate Biotin
Cross Reactivity Mouse
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Rat
Immunogen The immunogen for this antibody was NKG2D CD314.
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 22914