Please login to your online account to display your discounted pricing

Rat anti-NKG2D/CD314, Clone: CX5, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14353101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14353101

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Applications Flow Cytometry
Host Species Rat
Antigen NKG2D/CD314
Regulatory Status RUO
Immunogen The immunogen for this antibody was NKG2D CD314.
Clone CX5
Isotype IgG1, κ
Gene ID (Entrez) 22914
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Conjugate Biotin
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Format Affinity Purified
Concentration 0.5mg/mL
Purification Method Affinity Purified
Quantity 0.1mg