Please login to your online account to display your discounted pricing

Rat anti-Mouse IgM, Clone: II/41, PE-Cy5, Novus Biologicals™

$252.00 - $362.00

Specifications

Antigen Mouse IgM
Clone II/41
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was Phycoerythrin IgM.
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14306801

 
novus biologicals canada
NBP14306801MG
0.1mg Each for $362.00

NBP14306805

 
novus biologicals canada
NBP143068005MG
0.05mg Each for $252.00
Description & Specifications

Specifications

Antigen Mouse IgM
Clone II/41
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was Phycoerythrin IgM.
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate PE-Cy5
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry