missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-Mouse IgM, Clone: II/41, FITC, Novus Biologicals™

$252.00 - $725.00

Specifications

Antigen Mouse IgM
Clone II/41
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Fluorescein IgM.
Formulation Phosphate buffer pH 7.2
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14394705 Novus Biologicals Canada
NBP143947005MG
0.05mg Each for $252.00
NBP1439475 Novus Biologicals Canada
NBP14394705MG
0.5mg Each for $725.00
Specifications Description & Specifications

Specifications

Mouse IgM
0.5mg/mL
Phosphate buffer pH 7.2
Primary
Rat
FITC
Affinity Purified
II/41
The immunogen for this antibody was Fluorescein IgM.
Monoclonal
Store at 4°C. Do not freeze.
IgG2a, κ
Affinity Purified
Flow Cytometry