Please login to your online account to display your discounted pricing

Rat anti-Mouse IgM, Clone: eB121-15F9, Biotin, Secondary Antibody, Novus Biologicals™

$219.00 - $329.00

Specifications

Antigen IgM
Clone eB121-15F9
Immunogen The immunogen for this antibody was IgM.
Gene ID (Entrez) 3507
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14352701

 
novus biologicals canada
NBP14352701MG
0.1mg Each for $219.00

NBP1435275

 
novus biologicals canada
NBP14352705MG
0.5mg Each for $329.00
Description & Specifications

Specifications

Antigen IgM
Clone eB121-15F9
Immunogen The immunogen for this antibody was IgM.
Gene ID (Entrez) 3507
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Secondary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry