Please login to your online account to display your discounted pricing

Rat anti-Mouse IgM, Clone: eB121-15F9, Biotin, Secondary Antibody, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14352705MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1435275

  • / Each
 

Description & Specifications

Specifications

Immunogen The immunogen for this antibody was IgM.
Antigen IgM
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Rat
Applications Flow Cytometry
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 0.5mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Secondary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3507
Clone eB121-15F9
Conjugate Biotin
Cross Reactivity Mouse