Please login to your online account to display your discounted pricing

Rat anti-MHC Class II, Clone: M5/114.15.2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14331205MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1433125

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Rat
Immunogen The immunogen for this antibody was MHC Class II.
Cross Reactivity Mouse
Antigen MHC Class II
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Isotype IgG2b, κ
Quantity 0.5mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Conjugate Unlabeled
Purification Method Affinity Purified
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3108
Test Specificity The monoclonal antibody reacts with the mouse major histocompatibility complex class II, both I-A and I-E subregion-encoded glycoproteins (I-A b, I-A d, I-A q, I-E d, I-E k, not I-A f, I-A k, or I-A s). It detects a polymorphic determinant present on B cells, monocytes, macrophages, dendritic cells, and activated T lymphocytes from mice carrying the H-2 b, H-2 d, H-2 q, H-2 p, H-2 r and H-2 u but not from mice carrying the H-2 s or H-2 f haplotypes. The M5/114 mAb is reported to inhibit I-A-restricted T cell responses of the H-2 b, H-2 d, H-2 q, H-2 u but not H-2 f, H-2 k, or H-2 s haplotypes.
Clone M5/114.15.2
Concentration 0.5mg/mL
Primary or Secondary Primary