Please login to your online account to display your discounted pricing

Rat anti-MHC Class II, Clone: M5/114.15.2, APC, Novus Biologicals™

$252.00 - $362.00

Specifications

Antigen MHC Class II
Clone M5/114.15.2
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was MHC Class II.
Gene ID (Entrez) 3108
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14299701

 
novus biologicals canada
NBP14299701MG
0.1mg Each for $362.00

NBP14299705

 
novus biologicals canada
NBP142997005MG
0.05mg Each for $252.00
Description & Specifications

Specifications

Antigen MHC Class II
Clone M5/114.15.2
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was MHC Class II.
Gene ID (Entrez) 3108
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Rat
Isotype IgG2b, κ
Conjugate Allophycocyanin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry