Please login to your online account to display your discounted pricing

Rat anti-MAdCAM-1, Clone: MECA-367, Biotin, Novus Biologicals™

$252.00 - $725.00

Specifications

Antigen MAdCAM-1
Clone MECA-367
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was MAdCAM1.
Gene ID (Entrez) 8174
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14350001

 
novus biologicals canada
NBP14350001MG
0.1mg Each for $329.00

NBP14350005

 
novus biologicals canada
NBP143500005MG
0.05mg Each for $252.00

NBP1435005

 
novus biologicals canada
NBP14350005MG
0.5mg Each for $725.00
Description & Specifications

Specifications

Antigen MAdCAM-1
Clone MECA-367
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was MAdCAM1.
Gene ID (Entrez) 8174
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry