Please login to your online account to display your discounted pricing

Rat anti-M-CSF R, Clone: AFS98, Novus Biologicals™

$274.00 - $538.00

Specifications

Antigen M-CSF R
Clone AFS98
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD115.
Gene Accession No. P07333
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14336301

 
novus biologicals canada
NBP14336301MG
0.1mg Each for $329.00

NBP14336305

 
novus biologicals canada
NBP143363005MG
0.05mg Each for $274.00

NBP1433635

 
novus biologicals canada
NBP14336305MG
0.5mg Each for $538.00
Description & Specifications

Specifications

Antigen M-CSF R
Clone AFS98
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD115.
Gene Accession No. P07333
Gene ID (Entrez) 1436
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Blocking
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Neutralization
Applications Western Blotting