Please login to your online account to display your discounted pricing

Rat anti-M-CSF R, Clone: AFS98, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14336305MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1433635

  • / Each
 

Description & Specifications

Specifications

Applications Blocking
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Neutralization
Applications Western Blotting
Primary or Secondary Primary
Format Affinity Purified
Immunogen The immunogen for this antibody was CD115.
Formulation Phosphate buffer pH 7.2
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Quantity 0.5mg
Host Species Rat
Purification Method Affinity Purified
Isotype IgG2a, κ
Cross Reactivity Mouse
Antigen M-CSF R
Concentration 0.5mg/mL
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1436
Clone AFS98
Gene Accession No. P07333
Conjugate Unlabeled