Please login to your online account to display your discounted pricing

Rat anti-M-CSF R, Clone: 12-3A3-1B10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1433620025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14336203

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen The immunogen for this antibody was CD115.
Format Affinity Purified
Antigen M-CSF R
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 1436
Primary or Secondary Primary
Host Species Rat
Formulation Phosphate buffer pH 7.2
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Clone 12-3A3-1B10
Isotype IgG1
Monoclonal or Polyclonal Monoclonal
Quantity 0.025mg
Purification Method Affinity Purified