Please login to your online account to display your discounted pricing

Rat anti-M-CSF R, Clone: 12-3A3-1B10, Biotin, Novus Biologicals™

$175.00 - $329.00

Specifications

Antigen M-CSF R
Clone 12-3A3-1B10
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD115.
Gene ID (Entrez) 1436
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14359101

 
novus biologicals canada
NBP14359101MG
0.1mg Each for $329.00

NBP14359103

 
novus biologicals canada
NBP1435910025MG
0.025mg Each for $175.00
Description & Specifications

Specifications

Antigen M-CSF R
Clone 12-3A3-1B10
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD115.
Gene ID (Entrez) 1436
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG1
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry