Please login to your online account to display your discounted pricing

Rat anti-LYVE-1, Clone: ALY7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1434110025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14341103

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Format Affinity Purified
Cross Reactivity Human
Cross Reactivity Mouse
Immunogen The immunogen for this antibody was LYVE1.
Gene ID (Entrez) 10894
Primary or Secondary Primary
Purification Method Affinity Purified
Host Species Rat
Monoclonal or Polyclonal Monoclonal
Quantity 0.025mg
Isotype IgG1
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Concentration 0.5mg/mL
Clone ALY7
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Antigen LYVE-1
Test Specificity Human reactivity reported in scientific literature (PMID: 23975155)
Formulation Aqueous buffer (contains stabilizer if necessary