Please login to your online account to display your discounted pricing

Rat anti-LYVE-1, Clone: ALY7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14341101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14341101

  • / Each
 

Description & Specifications

Specifications

Antigen LYVE-1
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Clone ALY7
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation Aqueous buffer (contains stabilizer if necessary
Host Species Rat
Immunogen The immunogen for this antibody was LYVE1.
Isotype IgG1
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 10894
Test Specificity Human reactivity reported in scientific literature (PMID: 23975155)
Target Species Human
Target Species Murine