Please login to your online account to display your discounted pricing

Rat anti-LYVE-1, Clone: ALY7, Biotin, Novus Biologicals™

$175.00 - $329.00

Specifications

Antigen LYVE-1
Clone ALY7
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was LYVE1.
Gene ID (Entrez) 10894
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14364501

 
novus biologicals canada
NBP14364501MG
0.1mg Each for $329.00

NBP14364503

 
novus biologicals canada
NBP1436450025MG
0.025mg Each for $175.00
Description & Specifications

Specifications

Antigen LYVE-1
Clone ALY7
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was LYVE1.
Gene ID (Entrez) 10894
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG1
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)