Please login to your online account to display your discounted pricing

Rat anti-Lymphotoxin beta R/TNFRSF3, Clone: 3C8, Biotin, Novus Biologicals™

$175.00 - $362.00

Specifications

Antigen Lymphotoxin beta R/TNFRSF3
Clone 3C8
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Lymphotoxinbeta Receptor.
Gene ID (Entrez) 4055
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14355001

 
novus biologicals canada
NBP14355001MG
0.1mg Each for $362.00

NBP14355003

 
novus biologicals canada
NBP1435500025MG
0.025mg Each for $175.00
Description & Specifications

Specifications

Antigen Lymphotoxin beta R/TNFRSF3
Clone 3C8
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Lymphotoxinbeta Receptor.
Gene ID (Entrez) 4055
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG1, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry