Please login to your online account to display your discounted pricing

Rat anti-Ly-6G, Clone: RB6-8C5, PE, Novus Biologicals™

$406.00 - $406.00

Specifications

Antigen Ly-6G
Clone RB6-8C5
Concentration 0.2mg/mL
Immunogen Gr1).
Gene ID (Entrez) 546644
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20044010

 
novus biologicals canada
NBP200440100UG
100μg Each for $406.00
Description & Specifications

Specifications

Antigen Ly-6G
Clone RB6-8C5
Concentration 0.2mg/mL
Immunogen Gr1).
Gene ID (Entrez) 546644
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2b, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications In vivo Assay